Gene Transcripts
- Gene Symbol
- MIR18A
- Entrez Gene ID
- 406953
- Gene Name
- microRNA 18a
- Gene Aliases
- MIR18, MIRN18, MIRN18A, hsa-mir-18, hsa-mir-18a, miR-18, miRNA18A, mir-18a
- Chromosome Location
- Chr. 13: 91350751 - 91350821 on Build GRCh38
- UniGene ID
- Hs.24115
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
RNADB ID |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB176708.1 |
- |
LIT3364 oncomir-1 |
2 |
5046 |
|
View Full Product Details