Gene Transcripts
- Gene Symbol
- ALDOAP2
- Entrez Gene ID
- 228
- Gene Name
- aldolase, fructose-bisphosphate A pseudogene 2
- Gene Aliases
- Chromosome Location
- Chr. 10: 125666680 - 125668056 on Build GRCh38
- UniGene ID
- Hs.652473
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
RNADB ID |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M21191.1 |
- |
LIT2112 ALDOAP2 |
1 |
1950 |
|
View Full Product Details