Gene Transcripts

Gene Symbol
SRRM2
Entrez Gene ID
23524
Gene Name
serine/arginine repetitive matrix 2
Gene Aliases
300-KD, CWF21, Cwc21, HSPC075, SRL300, SRm300
Chromosome Location
Chr. 16: 2752329 - 2771412 on Build GRCh38
UniGene ID
Hs.433343
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006720874.3 XP_006720937.1 10 2151
GenBank mRNA BX537511.1 - 1 605

View Full Product Details