Gene Transcripts
- Gene Symbol
- AVPI1
- Entrez Gene ID
- 60370
- Gene Name
- arginine vasopressin induced 1
- Gene Aliases
- PP5395, VIP32, VIT32
- Chromosome Location
- Chr. 10: 97677422 - 97687432 on Build GRCh38
- UniGene ID
- Hs.23918
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BI520308.1 |
- |
1 |
34 |
|
Chr. 10 - 97679689 - 97687412 [-] |
GenBankmRNA |
DA063360.1 |
- |
1 |
23 |
|
Chr. 10 - 97686843 - 97687401 [-] |
GenBankmRNA |
BI755867.1 |
- |
1 |
12 |
|
Chr. 10 - 97679853 - 97687390 [-] |
View Full Product Details