Gene Transcripts
- Gene Symbol
- NREP
- Entrez Gene ID
- 9315
- Gene Name
- neuronal regeneration related protein
- Gene Aliases
- C5orf13, D4S114, P311, PRO1873, PTZ17, SEZ17
- Chromosome Location
- Chr. 5: 111729303 - 111976931 on Build GRCh38
- UniGene ID
- Hs.36053
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK021624.1 |
- |
1 |
185 |
|
AK310585.1 |
- |
2 |
429 |
|
View Full Product Details