Gene Transcripts
- Gene Symbol
- RRP12
- Entrez Gene ID
- 23223
- Gene Name
- ribosomal RNA processing 12 homolog
- Gene Aliases
- KIAA0690
- Chromosome Location
- Chr. 10: 97356701 - 97401370 on Build GRCh38
- UniGene ID
- Hs.434251
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC042908.1 |
- |
1 |
802 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM914492.1 |
- |
1 |
803 |
|
Chr. 10 - 97393044 - 97393987 [-] |
View Full Product Details