Gene Transcripts
- Gene Symbol
- RIN3
- Entrez Gene ID
- 79890
- Gene Name
- Ras and Rab interactor 3
- Gene Aliases
- Chromosome Location
- Chr. 14: 92513773 - 92688994 on Build GRCh38
- UniGene ID
- Hs.326822
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK096145.1 |
- |
1 |
1979 |
|
BC034153.1 |
- |
1 |
783 |
|
View Full Product Details