Gene Transcripts
- Gene Symbol
- RASAL2
- Entrez Gene ID
- 9462
- Gene Name
- RAS protein activator like 2
- Gene Aliases
- NGAP
- Chromosome Location
- Chr. 1: 178093729 - 178479514 on Build GRCh38
- UniGene ID
- Hs.496139
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK000981.1 |
- |
1 |
653 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AU118091.1 |
- |
1 |
653 |
|
Chr. 1 - 178143886 - 178144723 [+] |
View Full Product Details