Gene Transcripts
- Gene Symbol
- GAB1
- Entrez Gene ID
- 2549
- Gene Name
- GRB2 associated binding protein 1
- Gene Aliases
- Chromosome Location
- Chr. 4: 143336830 - 143474565 on Build GRCh38
- UniGene ID
- Hs.618456
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AF074997.1 |
- |
1 |
515 |
|
Chr. 4 - 143353050 - 143353673 [+] |
GenBankmRNA |
BX103277.1 |
- |
1 |
514 |
|
Chr. 4 - 143353050 - 143353687 [+] |
View Full Product Details