Gene Transcripts
- Gene Symbol
- KIN
- Entrez Gene ID
- 22944
- Gene Name
- Kin17 DNA and RNA binding protein
- Gene Aliases
- BTCD, KIN17, Rts2
- Chromosome Location
- Chr. 10: 7750962 - 7788027 on Build GRCh38
- UniGene ID
- Hs.397918
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CK003350.1 |
- |
1 |
647 |
|
Chr. 10 - 7751049 - 7751721 [-] |
GenBankmRNA |
BM782457.1 |
- |
1 |
422 |
|
Chr. 10 - 7750971 - 7751496 [-] |
View Full Product Details