Gene Transcripts
- Gene Symbol
- C5orf56
- Entrez Gene ID
- 441108
- Gene Name
- chromosome 5 open reading frame 56
- Gene Aliases
- Chromosome Location
- Chr. 5: 132410773 - 132476044 on Build GRCh38
- UniGene ID
- Hs.658288
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK025221.1 |
- |
1 |
108 |
|
View Full Product Details