Gene Transcripts

Gene Symbol
METTL15
Entrez Gene ID
196074
Gene Name
methyltransferase like 15
Gene Aliases
METT5D1
Chromosome Location
Chr. 11: 28108251 - 28527041 on Build GRCh38
UniGene ID
Hs.243326
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011519935.2 XP_011518237.1 5 905
XM_011519937.2 XP_011518239.1 2 146
XM_017017298.1 XP_016872787.1 6 944
XM_017017299.1 XP_016872788.1 2 1433
GenBank mRNA BC105287.1 - 5 599


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AK091261.1 - 1 2126 Chr. 11 - 28123518 - 28125860 [+]

View Full Product Details