Gene Transcripts
- Gene Symbol
- SYTL2
- Entrez Gene ID
- 54843
- Gene Name
- synaptotagmin like 2
- Gene Aliases
- CHR11SYT, EXO4, PPP1R151, SGA72M, SLP2, SLP2A
- Chromosome Location
- Chr. 11: 85694221 - 85811182 on Build GRCh38
- UniGene ID
- Hs.369520
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK056722.1 |
- |
1 |
833 |
|
View Full Product Details