Gene Transcripts
- Gene Symbol
- DNAI1
- Entrez Gene ID
- 27019
- Gene Name
- dynein axonemal intermediate chain 1
- Gene Aliases
- CILD1, DIC1, ICS1, PCD
- Chromosome Location
- Chr. 9: 34458752 - 34520989 on Build GRCh38
- UniGene ID
- Hs.112667
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL117535.1 |
- |
1 |
579 |
|
View Full Product Details