Gene Transcripts

Gene Symbol
C16orf72
Entrez Gene ID
29035
Gene Name
chromosome 16 open reading frame 72
Gene Aliases
PRO0149
Chromosome Location
Chr. 16: 9091329 - 9119698 on Build GRCh38
UniGene ID
Hs.221497
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011522462.2 XP_011520764.1 4 4398
GenBank mRNA AK057657.1 - 1 1762
AK095835.1 - 1 3127
AK123562.1 - 1 2626

View Full Product Details