Gene Transcripts
- Gene Symbol
- MLEC
- Entrez Gene ID
- 9761
- Gene Name
- malectin
- Gene Aliases
- KIAA0152
- Chromosome Location
- Chr. 12: 120687124 - 120701864 on Build GRCh38
- UniGene ID
- Hs.744910
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CN313738.1 |
- |
1 |
344 |
|
Chr. 12 - 120691411 - 120692019 [+] |
GenBankmRNA |
CN313740.1 |
- |
1 |
459 |
|
Chr. 12 - 120691302 - 120692080 [+] |
View Full Product Details