Gene Transcripts
- Gene Symbol
- LOC105379704
- Entrez Gene ID
- 105379704
- Gene Name
- CASP-like protein 4A1
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.606356
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC036479.1 |
- |
1 |
2059 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BI916626.1 |
- |
|
143 |
|
Chr. - - [-] |
View Full Product Details