Gene Transcripts

Gene Symbol
SNHG5
Entrez Gene ID
387066
Gene Name
small nucleolar RNA host gene 5
Gene Aliases
C6orf160, LINC00044, NCRNA00044, U50HG, bA33E24.2
Chromosome Location
Chr. 6: 85677007 - 85678733 on Build GRCh38
UniGene ID
Hs.292457
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK311192.1 - 1 65
BC009220.2 - 1 34
BC108291.1 - 1 52


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CD672298.1 - 1 54 Chr. 6 - 85677470 - 85678722 [-]
GenBankmRNA AL525144.3 - 1 57 Chr. 6 - 85677103 - 85678710 [-]
GenBankmRNA BM728819.1 - 1 53 Chr. 6 - 85677811 - 85678721 [-]
GenBankmRNA AB017710.3 - 1 692 Chr. 6 - 85677007 - 85679361 [-]
GenBankmRNA BG472221.1 - 54 Chr. - - [-]
GenBankmRNA BG697433.1 - 1 52 Chr. 6 - 85677426 - 85678720 [-]

View Full Product Details