Gene Transcripts

Gene Symbol
AP3S1
Entrez Gene ID
1176
Gene Name
adaptor related protein complex 3 sigma 1 subunit
Gene Aliases
CLAPS3, Sigma3A
Chromosome Location
Chr. 5: 115841606 - 115914081 on Build GRCh38
UniGene ID
Hs.406191
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017009024.1 XP_016864513.1 7 1507
GenBank mRNA AK124911.1 - 1 894
AK126876.1 - 1 3544

View Full Product Details