Gene Transcripts
- Gene Symbol
- PDCD6
- Entrez Gene ID
- 10016
- Gene Name
- programmed cell death 6
- Gene Aliases
- ALG-2, ALG2, PEF1B
- Chromosome Location
- Chr. 5: 271621 - 314974 on Build GRCh38
- UniGene ID
- Hs.50823
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK124302.1 |
- |
1 |
1540 |
|
AK128087.1 |
- |
|
3166 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK124667.1 |
- |
1 |
654 |
|
Chr. 5 - 310457 - 312809 [+] |
View Full Product Details