Gene Transcripts
- Gene Symbol
- SLC13A3
- Entrez Gene ID
- 64849
- Gene Name
- solute carrier family 13 member 3
- Gene Aliases
- NADC3, SDCT2
- Chromosome Location
- Chr. 20: 46557823 - 46684485 on Build GRCh38
- UniGene ID
- Hs.655498
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK023307.1 |
- |
1 |
314 |
|
View Full Product Details