Gene Transcripts
- Gene Symbol
- RPS6P6
- Entrez Gene ID
- 728131
- Gene Name
- ribosomal protein S6 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 4: 82494806 - 82495934 on Build GRCh38
- UniGene ID
- Hs.507676
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC052613.1 |
- |
1 |
762 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BQ711654.1 |
- |
1 |
774 |
|
Chr. 4 - 82495363 - 82496279 [-] |
View Full Product Details