Gene Transcripts

Gene Symbol
TECTA
Entrez Gene ID
7007
Gene Name
tectorin alpha
Gene Aliases
DFNA12, DFNA8, DFNB21
Chromosome Location
Chr. 11: 121102666 - 121190806 on Build GRCh38
UniGene ID
Hs.248162
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF143331.1 - 1 518


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CR743857.1 - 1 504 Chr. 11 - 121190774 - 121191486 [+]
GenBankmRNA BX110584.1 - 1 504 Chr. 11 - 121190774 - 121191486 [+]

View Full Product Details