Gene Transcripts
- Gene Symbol
- Slc38a6
- Entrez Gene ID
- 625098
- Gene Name
- solute carrier family 38, member 6
- Gene Aliases
- AW322671, EG625098
- Chromosome Location
- Chr. 12: 73286794 - 73361998 on Build GRCm38
- UniGene ID
- Mm.32075
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK141055.1 |
- |
2 |
1501 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AA165791.1 |
- |
1 |
448 |
|
Chr. 12 - 73360788 - 73361303 [+] |
View Full Product Details