Gene Transcripts
- Gene Symbol
- Smo
- Entrez Gene ID
- 319757
- Gene Name
- smoothened homolog (Drosophila)
- Gene Aliases
- E130215L21Rik, Smoh, bnb, smoothened
- Chromosome Location
- Chr. 6: 29735497 - 29761366 on Build GRCm38
- UniGene ID
- Mm.29279
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK052407.1 |
- |
|
776 |
|
View Full Product Details