Gene Transcripts
- Gene Symbol
- Pabpc1l
- Entrez Gene ID
- 381404
- Gene Name
- poly(A) binding protein, cytoplasmic 1-like
- Gene Aliases
- 1810053B01Rik, Epab
- Chromosome Location
- Chr. 2: 164024770 - 164050538 on Build GRCm38
- UniGene ID
- Mm.82301
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK007853.1 |
- |
1 |
20 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY707856.1 |
- |
1 |
18 |
|
Chr. 2 - 164047388 - 164050479 [+] |
View Full Product Details