Gene Transcripts
- Gene Symbol
- Pim1
- Entrez Gene ID
- 18712
- Gene Name
- proviral integration site 1
- Gene Aliases
- Pim-1
- Chromosome Location
- Chr. 17: 29491045 - 29495465 on Build GRCm38
- UniGene ID
- Mm.405293
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK047301.1 |
- |
|
1648 |
|
View Full Product Details