Gene Transcripts

Gene Symbol
Sike1
Entrez Gene ID
66641
Gene Name
suppressor of IKBKE 1
Gene Aliases
2810005O12Rik, 5730470L24Rik, AI450236, AI839862, Sike, Sikeb
Chromosome Location
Chr. 3: 102995740 - 103003914 on Build GRCm38
UniGene ID
Mm.208619
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK139055.1 - 1757


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CF551415.1 - 1 728 Chr. 3 - 103007165 - 103008013 [+]
GenBankmRNA AA185042.1 - 1 171 Chr. 3 - 103007719 - 103008182 [+]
GenBankmRNA BQ572892.1 - 1 548 Chr. 3 - 103007343 - 103008101 [+]
GenBankmRNA AI646656.1 - 1 172 Chr. 3 - 103007719 - 103008171 [+]

View Full Product Details