Gene Transcripts
- Gene Symbol
- Lrrc9
- Entrez Gene ID
- 78257
- Gene Name
- leucine rich repeat containing 9
- Gene Aliases
- 4921529O18Rik, 4930432K16Rik
- Chromosome Location
- Chr. 12: 72433940 - 72530174 on Build GRCm38
- UniGene ID
- Mm.266280
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK144081.1 |
- |
|
257 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CJ068761.1 |
- |
2 |
257 |
|
Chr. 12 - 72520091 - 72530575 [+] |
View Full Product Details