Gene Transcripts
- Gene Symbol
- Atxn7l3b
- Entrez Gene ID
- 382423
- Gene Name
- ataxin 7-like 3B
- Gene Aliases
- 4921506J03Rik, 6230409E21Rik, AI315132, ENSMUSG00000074747
- Chromosome Location
- Chr. 10: 112925428 - 112929026 on Build GRCm38
- UniGene ID
- Mm.431239
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK018082.1 |
- |
|
781 |
|
View Full Product Details