Gene Transcripts
- Gene Symbol
- Cbx1
- Entrez Gene ID
- 12412
- Gene Name
- chromobox 1
- Gene Aliases
- Cbx, Cbx-rs2, E430007M08Rik, HP1B, Hp1beta, M31, MOD1
- Chromosome Location
- Chr. 11: 96788679 - 96808982 on Build GRCm38
- UniGene ID
- Mm.29055
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BE625817.1 |
- |
1 |
56 |
|
Chr. 11 - 96807224 - 96807677 [+] |
GenBankmRNA |
BB637650.1 |
- |
1 |
79 |
|
Chr. 11 - 96807201 - 96807817 [+] |
GenBankmRNA |
BY746258.1 |
- |
|
68 |
|
Chr. - - [-] |
GenBankmRNA |
CK621202.1 |
- |
1 |
51 |
|
Chr. 11 - 96807229 - 96807761 [+] |
GenBankmRNA |
AW985572.1 |
- |
1 |
84 |
|
Chr. 11 - 96807210 - 96807649 [+] |
GenBankmRNA |
CK790554.1 |
- |
1 |
42 |
|
Chr. 11 - 96807251 - 96807959 [+] |
View Full Product Details