Gene Transcripts
- Gene Symbol
- Wdr27
- Entrez Gene ID
- 71682
- Gene Name
- WD repeat domain 27
- Gene Aliases
- 0610012K18Rik
- Chromosome Location
- Chr. 17: 14784832 - 14943151 on Build GRCm38
- UniGene ID
- Mm.122698
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK084217.1 |
- |
2 |
1640 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB466570.2 |
- |
1 |
543 |
|
Chr. 17 - 14937843 - 14938520 [-] |
View Full Product Details