Gene Transcripts
- Gene Symbol
- Asb13
- Entrez Gene ID
- 142688
- Gene Name
- ankyrin repeat and SOCS box-containing 13
- Gene Aliases
- 2210015B19Rik, 6430573K02Rik, Asb-13, C85285
- Chromosome Location
- Chr. 13: 3634012 - 3651779 on Build GRCm38
- UniGene ID
- Mm.253061
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK008728.1 |
- |
2 |
136 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY708630.1 |
- |
2 |
133 |
|
Chr. 13 - 3634089 - 3634782 [+] |
View Full Product Details