Gene Transcripts

Gene Symbol
Asb13
Entrez Gene ID
142688
Gene Name
ankyrin repeat and SOCS box-containing 13
Gene Aliases
2210015B19Rik, 6430573K02Rik, Asb-13, C85285
Chromosome Location
Chr. 13: 3634012 - 3651779 on Build GRCm38
UniGene ID
Mm.253061
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK008728.1 - 2 166


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BY708630.1 - 2 163 Chr. 13 - 3634089 - 3634782 [+]

View Full Product Details