Gene Transcripts
- Gene Symbol
- IGF2BP2-AS1
- Entrez Gene ID
- 646600
- Gene Name
- IGF2BP2 antisense RNA 1
- Gene Aliases
- C3orf65
- Chromosome Location
- Chr. 3: 185713252 - 185729787 on Build GRCh38
- UniGene ID
- Hs.647949
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_126326.1 |
|
4 |
913 |
|
GenBank mRNA |
BC039525.1 |
- |
2 |
187 |
|
BC041450.1 |
- |
4 |
912 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000456447.5 |
- |
4 |
913 |
|
Chr. 3 - 185713252 - 185729787 [+] |
GENCODE |
ENST00000496717.1 |
- |
2 |
187 |
|
Chr. 3 - 185722464 - 185729787 [+] |
View Full Product Details