Gene Transcripts
- Gene Symbol
- COLCA1
- Entrez Gene ID
- 399948
- Gene Name
- colorectal cancer associated 1
- Gene Aliases
- C11orf92, CASC12, LOH11CR1F
- Chromosome Location
- Chr. 11: 111293389 - 111305048 on Build GRCh38
- UniGene ID
- Hs.729225
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000532918.4 |
- |
2 |
2572 |
|
Chr. 11 - 111293389 - 111305045 [-] |
GENCODE |
ENST00000355430.4 |
- |
2 |
2705 |
|
Chr. 11 - 111293446 - 111305045 [-] |
GENCODE |
ENST00000620864.1 |
- |
|
2702 |
|
Chr. - - [-] |
GENCODE |
ENST00000540738.3 |
- |
|
2743 |
|
Chr. 11 - 111296104 - 111296478 [-] |
View Full Product Details