Gene Transcripts

Gene Symbol
C16orf47
Entrez Gene ID
388289
Gene Name
chromosome 16 open reading frame 47
Gene Aliases
Chromosome Location
Chr. 16: 73125865 - 73812449 on Build GRCh38
UniGene ID
Hs.371893
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207385.1 NP_997268.1 4 1261
XM_017023228.1 XP_016878717.1 9 2943
XM_017023229.1 XP_016878718.1 4 1549
GenBank mRNA AK129695.1 - 4 1261

View Full Product Details