Gene Transcripts
- Gene Symbol
- NTM-AS1
- Entrez Gene ID
- 101929637
- Gene Name
- NTM antisense RNA 1
- Gene Aliases
- C11orf39
- Chromosome Location
- Chr. 11: 131662129 - 131663556 on Build GRCh38
- UniGene ID
- Hs.730330
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_126159.1 |
|
1 |
133 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000416725.1 |
- |
1 |
160 |
|
Chr. 11 - 131662131 - 131663583 [-] |
View Full Product Details