Gene Transcripts

Gene Symbol
SMARCA5-AS1
Entrez Gene ID
100128055
Gene Name
SMARCA5 antisense RNA 1
Gene Aliases
Chromosome Location
Chr. 4: 143513472 - 143514837 on Build GRCh38
UniGene ID
Hs.707570
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_104027.1 1 367
GenBank mRNA AK307172.1 - 1 137
HY014185.1 - 1 367


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DA405791.1 - 1 137 Chr. 4 - 143513823 - 143514607 [-]
GENCODE ENST00000500800.2 - 1 165 Chr. 4 - 143513472 - 143514635 [-]
GenBankmRNA DA333970.1 - 1 165 Chr. 4 - 143513892 - 143514635 [-]

View Full Product Details