Gene Transcripts

Gene Symbol
Vmn1r53
Entrez Gene ID
113853
Gene Name
vomeronasal 1 receptor 53
Gene Aliases
V1rb3, VN5
Chromosome Location
Chr. 6: 90223317 - 90224438 on Build GRCm38
UniGene ID
Mm.434332
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053226.2 NP_444456.2 1 631
GenBank mRNA BC125605.1 - 1 631
BC132002.1 - 1 631 40130279

View Full Product Details