Gene Transcripts

Gene Symbol
Sned1
Entrez Gene ID
208777
Gene Name
sushi, nidogen and EGF-like domains 1
Gene Aliases
6720455I24Rik, AI197264, AI642697, D430044C15Rik, SST3, Snep
Chromosome Location
Chr. 1: 93235815 - 93301065 on Build GRCm38
UniGene ID
Mm.271897
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172463.4 NP_766051.4 21 3022
GenBank mRNA AJ584850.1 - 21 3078
AY169783.1 - 21 3077
BC145886.1 - 21 3022 40131189

View Full Product Details