Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001242339.1 | NP_001229268.1 | 10 | 1029 | |
NM_001323067.1 | NP_001309996.1 | 8 | 768 | ||
NM_001323068.1 | NP_001309997.1 | 8 | 905 | ||
NM_001323069.1 | NP_001309998.1 | 8 | 900 | ||
NM_001323070.1 | NP_001309999.1 | 3 | 715 | ||
NM_001323071.1 | NP_001310000.1 | 9 | 1021 | ||
NM_001323072.1 | NP_001310001.1 | 9 | 1128 | ||
NM_001323073.1 | NP_001310002.1 | 4 | 389 | ||
NM_001323074.1 | NP_001310003.1 | 3 | 715 | ||
NM_002627.4 | NP_002618.1 | 8 | 905 | ||
XM_005252465.4 | XP_005252522.1 | 8 | 950 | ||
XM_005252466.4 | XP_005252523.1 | 8 | 950 | ||
XM_005252467.2 | XP_005252524.1 | 8 | 874 | ||
XM_006717449.1 | XP_006717512.1 | 8 | 874 | ||
GenBank mRNA | AK092597.1 | - | 10 | 1029 | |
AK129843.1 | - | 444 | |||
AK291841.1 | - | 8 | 877 | ||
AK308003.1 | - | 8 | 868 | ||
AK308226.1 | - | 4 | 305 | ||
BC002536.1 | - | 8 | 865 | 3140233 | |
BC029138.1 | - | 8 | 905 | 5180268 | |
D25328.1 | - | 8 | 862 |