Gene Transcripts

Gene Symbol
Olfr11
Entrez Gene ID
218066
Gene Name
olfactory receptor 11
Gene Aliases
M49, MOR256-11, ORL121
Chromosome Location
Chr. 13: 21638580 - 21639521 on Build GRCm38
UniGene ID
Mm.484775
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146542.2 NP_666753.2 1 728
GenBank mRNA BC111579.1 - 1 726

View Full Product Details