Gene Transcripts

Gene Symbol
Nrsn2
Entrez Gene ID
228777
Gene Name
neurensin 2
Gene Aliases
Gm123, Neurensin-2
Chromosome Location
Chr. 2: 152368755 - 152377153 on Build GRCm38
UniGene ID
Mm.330667
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009948.1 NP_001009948.1 3 300
GenBank mRNA AK140190.1 - 3 317
BC088982.1 - 3 332 5700110

View Full Product Details