Gene Transcripts
- Gene Symbol
- Zfyve9
- Entrez Gene ID
- 230597
- Gene Name
- zinc finger, FYVE domain containing 9
- Gene Aliases
- E030027L17, Madhip, NSP, SARA, SMADIP
- Chromosome Location
- Chr. 4: 108637834 - 108781148 on Build GRCm38
- UniGene ID
- Mm.196776
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_183300.2 |
NP_899123.2
|
8, 9 |
1135 |
|
GenBank mRNA |
DQ303413.1 |
- |
8, 9 |
1135 |
|
View Full Product Details