Gene Transcripts

Gene Symbol
Slc38a8
Entrez Gene ID
234788
Gene Name
solute carrier family 38, member 8
Gene Aliases
Gm587
Chromosome Location
Chr. 8: 119479574 - 119501708 on Build GRCm38
UniGene ID
Mm.302096
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009950.1 NP_001009950.1 8 1020
GenBank mRNA BC089013.1 - 8 1020

View Full Product Details