Gene Transcripts

Gene Symbol
POU3F4
Entrez Gene ID
5456
Gene Name
POU class 3 homeobox 4
Gene Aliases
BRAIN-4, BRN-4, BRN4, DFN3, DFNX2, OCT-9, OTF-9, OTF9
Chromosome Location
Chr. X: 83508261 - 83509767 on Build GRCh38
UniGene ID
Hs.2229
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000307.4 NP_000298.3 1 1047
GenBank mRNA AK314967.1 - 1 1047
X82324.1 - 1 1015

View Full Product Details