Gene Transcripts

Gene Symbol
Slfn14
Entrez Gene ID
237890
Gene Name
schlafen 14
Gene Aliases
Gm20, Slfn14-ps
Chromosome Location
Chr. 11: 83275110 - 83286726 on Build GRCm38
UniGene ID
Mm.297670
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166028.1 NP_001159500.1 4 1345

View Full Product Details