Gene Transcripts

Gene Symbol
Otop2
Entrez Gene ID
237987
Gene Name
otopetrin 2
Gene Aliases
4732464P15
Chromosome Location
Chr. 11: 115307163 - 115332306 on Build GRCm38
UniGene ID
Mm.44542
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172801.2 NP_766389.2 7 1317
GenBank mRNA AB087504.1 - 6 1202
AK028866.1 - 7 1318

View Full Product Details