Gene Transcripts

Gene Symbol
Akap5
Entrez Gene ID
238276
Gene Name
A kinase (PRKA) anchor protein 5
Gene Aliases
3526401B18Rik, AKAP 150, AKAP-5, AKAP150, BB098886, Gm258, P150
Chromosome Location
Chr. 12: 76324402 - 76334151 on Build GRCm38
UniGene ID
Mm.311452
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101471.1 NP_001094941.1 2 516

View Full Product Details